2,6-Dibromonaphthalene ≥99%
Leverantör: Thermo Scientific
L16134.03 L16134.06 L16134.14
L16134.03EA 1575 SEK
L16134.03 L16134.06 L16134.14
2,6-Dibromonaphthalene ≥99%
2,6-Dibromonaphthalene
Purity: 99% CAS No.: 13720-06-4 Molecular Formula: C10H6Br2 Formula Weight: 285.97 Melting Point: 162-167° MDL No.: MFCD01026462
Formel: C₁₀H₆Br₂
MW: 285,97 g/mol
Smältpunkt: 162…167 °C
Storage Temperature: Omgivande
MDL Number: MFCD01026462
CAS nummer: 13720-06-4