1,1,3,3-Tetramethyl-2-thiourea 98
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: N,N,N',N'-Tetramethylthiourea, Tetramethyl-2-thiourea, Tetramethylthiourea
L13392.06 L13392.14
L13392.06EA 565 SEK
L13392.06 L13392.14
1,1,3,3-Tetramethyl-2-thiourea 98
1,1,3,3-Tetramethyl-2-thiourea
Purity: 98% CAS No.: 2782-91-4 Molecular Formula: C5H12N2S Formula Weight: 132.23 Flash Point: 190°(374°F) Odour: Odourless Boiling Point: 245° Beilstein Reference: 1744916 MDL No.: MFCD00008324
Formel: C₅H₁₂N₂S
MW: 132.23 g/mol
Kokpunkt: 245 °C
Smältpunkt: 75…77 °C
Flampunkt: 190 °C (374 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008324
CAS nummer: 2782-91-4
EINECS: 220-488-0