3-Cyanophenyl isothiocyanate ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Cyano-3-isothiocyanatobenzene, 3-Isothiocyanatobenzonitrile
L13045.03 L13045.06
L13045.03EA 1150 SEK
L13045.03 L13045.06
3-Cyanophenyl isothiocyanate ≥97%
3-Cyanophenyl isothiocyanate
Purity: 97% CAS No.: 3125-78-8 Molecular Formula: C8H4N2S Formula Weight: 160.20 Odour: Irritating Melting Point: 64-66° Beilstein Reference: 2803099 UN No.: 2923 MDL No.: MFCD00041070
Formel: C₈H₄N₂S
MW: 160,2 g/mol
Kokpunkt: 294 °C (1013 hPa)
Smältpunkt: 64…66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041070
CAS nummer: 3125-78-8
UN: 2923
ADR: 8,III