5-Hexenoic acid ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 5-Hexenoicacid
L12863.03 L12863.06 L12863.22
L12863.03EA 1475 SEK
L12863.03 L12863.06 L12863.22
5-Hexenoic acid ≥99%
5-Hexenoic acid
Purity: 99% CAS No.: 1577-22-6 Molecular Formula: C6H10O2 Formula Weight: 114.14 Flash Point: 104°(219°F) Refractive Index: 1.4340 Boiling Point: 107°/17mm Beilstein Reference: 1743172 UN No.: 3265 MDL No.: MFCD00046558
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 107 °C (17 mmHg)
Smältpunkt: –37 °C
Densitet: 0,961
Flampunkt: 104 °C (219 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00046558
CAS nummer: 1577-22-6
UN: 3265
ADR: 8,III