1-Bromo-3-phenoxypropane ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Bromopropyl phenyl ether, (3-Bromopropoxy)benzene, 3-Phenoxypropyl bromide
L12494.14 L12494.22
L12494.14EA 1650 SEK
L12494.14 L12494.22
1-Bromo-3-phenoxypropane ≥98%
1-Bromo-3-phenoxypropane
Purity: 98% CAS No.: 588-63-6 Molecular Formula: C6H5OCH2CH2CH2Br Formula Weight: 215.10 Flash Point: 96°(204°F) Refractive Index: 1.5460 Melting Point: 10-11° Boiling Point: 130-134°/14mm Beilstein Reference: 508978 MDL No.: MFCD00000256
Formel: (C₆H₅)OCH₂CH₂CH₂Br
Kokpunkt: 130…134 °C (14 mmHg)
Smältpunkt: 10…11 °C
Densitet: 1,365
Flampunkt: 96 °C (204 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000256
CAS nummer: 588-63-6
EINECS: 209-623-4