3-Hydroxy-p-anisidine 98%
Leverantör: Thermo Fisher Scientific
Fara
L10234.14 L10234.06
L10234.14EA 5075 SEK
L10234.14 L10234.06
3-Hydroxy-p-anisidine 98%
3-Hydroxy-p-anisidine
Purity: 98% CAS No.: 1687-53-2 Molecular Formula: C7H9NO2 Formula Weight: 139.16 Melting Point: 130-132° Beilstein Reference: 2717084
Formel: C₇H₉NO₂
Kokpunkt: 277 °C (1013 hPa)
Smältpunkt: 130…132 °C
Densitet: 1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010222
CAS nummer: 1687-53-2
EINECS: 216-873-8