Methyl-3,4-dihydroxybenzoate ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,4-Dihydroxybenzoic acid methyl ester, Methyl 3,4-dihydroxybenzoate
L09552.09 L09552.04
L09552.09EA 3700 SEK
L09552.09 L09552.04
Methyl-3,4-dihydroxybenzoate ≥97%
Methyl-3,4-dihydroxybenzoate
Purity: 97% CAS No.: 2150-43-8 Molecular Formula: C8H8O4 Formula Weight: 168.15 Odour: Odourless Melting Point: 134-138° Beilstein Reference: 1637939 MDL No.: MFCD00016437
Formel: C₈H₈O₄
MW: 168,15 g/mol
Smältpunkt: 134…138 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016437
CAS nummer: 2150-43-8