Ricinolja
Leverantör: Thermo Scientific
Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
L04224.AK L04224.AU L04224.0F
L04224.AKEA 655 SEK
L04224.AK L04224.AU L04224.0F
Ricinolja
Ricinolja
CAS No.: 8001-79-4 Flash Point: 230°(446°F) Refractive Index: 1.4780 Boiling Point: 313° Merck Reference: 14,1898 Optical Rotation: +5° (c=5 in ethanol)
Kokpunkt: 313 °C
Smältpunkt: –10 °C
Densitet: 0,961
Flampunkt: 230 °C (446 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00130746
CAS nummer: 8001-79-4
EINECS: 232-293-8
Merck Index: 14,1898