Tris(pentafluorophenyl)phosphine ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Perfluorotriphenylphosphine, Tris(perfluorophenyl)phosphine
L02748.03 L02748.06
L02748.03EA 1400 SEK
L02748.03 L02748.06
Tris(pentafluorophenyl)phosphine ≥97%
Tris(pentafluorophenyl)phosphine
Purity: 97% CAS No.: 1259-35-4 Molecular Formula: C18F15P Formula Weight: 532.15 Odour: Odourless Melting Point: 115-119° Beilstein Reference: 773243 MDL No.: MFCD00079654
Formel: C₁₈F₁₅P
MW: 532,15 g/mol
Smältpunkt: 115…119 °C
Storage Temperature: Omgivande
MDL Number: MFCD00079654
CAS nummer: 1259-35-4
EINECS: 215-021-2