3-(2-Hydroxyethyl)indole ≥97%
Leverantör: Thermo Fisher Scientific
Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
L02555.03 L02555.06
L02555.03EA 600 SEK
L02555.03 L02555.06
3-(2-Hydroxyethyl)indole ≥97%
3-(2-Hydroxyethyl)indole
Purity: 97% CAS No.: 526-55-6 Molecular Formula: C10H11NO Formula Weight: 161.20 Flash Point: 255°(491°F) Melting Point: 56-59° Boiling Point: 174°/2mm Beilstein Reference: 125553 Merck Reference: 14,9798 MDL No.: MFCD00005659
Formel: C₁₀H₁₁NO
Kokpunkt: 174 °C (2 mmHg)
Smältpunkt: 56…59 °C
Flampunkt: 255 °C (491 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005659
CAS nummer: 526-55-6
EINECS: 208-393-2
Merck Index: 14,9798