4-(Dimethylamino)pyridine 99%, prilled
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine
H51715.06 H51715.14 H51715.22
H51715.06EA 630 SEK
H51715.06 H51715.14 H51715.22
4-(Dimethylamino)pyridine 99%, prilled
4-(Dimethylamino)pyridine
Purity: 99% CAS No.: 1122-58-3 Molecular Formula: C7H10N2 Formula Weight: 122.17 Flash Point: 124°(255°F) Melting Point: 110-113° Boiling Point: 162°/50mm Beilstein Reference: 110354 UN No.: 2928 MDL No.: MFCD00006418
Formel: C₇H₁₀N₂
MW: 122.17 g/mol
Kokpunkt: 162 °C (50 mmHg)
Smältpunkt: 110…113 °C
Flampunkt: 124 °C (255 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006418
CAS nummer: 1122-58-3
EINECS: 214-353-5
UN: 2928
ADR: 6.1 I