3-Fluoro-2-hydroxypyridine ≥97%
Leverantör: Thermo Scientific
Varning
H31705.MD H31705.03 H31705.06
H31705.MDEA 2425 SEK
H31705.MD H31705.03 H31705.06
3-Fluoro-2-hydroxypyridine ≥97%
3-Fluoro-2-hydroxypyridine
Purity: 97% CAS No.: 1547-29-1 Molecular Formula: C5H4FNO Formula Weight: 113.09 Odour: Odourless Melting Point: 157-161° MDL No.: MFCD03095042
Formel: C₅H₄FNO
MW: 113,09 g/mol
Smältpunkt: 157…161 °C
Storage Temperature: Omgivande
MDL Number: MFCD03095042
CAS nummer: 1547-29-1