4-Mercapto-4-methyl-2-pentanone ≥98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
Purity: 98%
CAS No.: 19872-52-7
Molecular Formula: C6H12SO
Formula Weight: 132.23
Storage Temperature: -30°C to -10°C
Flash Point: 54°(129°F)
Refractive Index: 1.4620
Odour: Unpleasant
Boiling Point: 174°
UN No.: 3336
MDL No.: MFCD00085208
Formel:
C₆H₁₂OS Kokpunkt: 174 °C Densitet: 0,961 Flampunkt: 54 °C (129 °F) |
MDL Number:
MFCD00085208 CAS nummer: 19872-52-7 EINECS: 243-386-8 ADR: 3,III |