1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Butyl-2,3-dimethyl-1H-imidazol-3-ium hexafluorophosphate(V)
H27827.06 H27827.18
H27827.06EA 1300 SEK
H27827.06 H27827.18
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate ≥99%
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate
Purity: 99% CAS No.: 227617-70-1 Molecular Formula: C9H17F6N2P Formula Weight: 298.21 Refractive Index: 1.423 Melting Point: 48-50° MDL No.: MFCD03790877
Formel: C₉H₁₇F₆N₂P
MW: 298,21 g/mol
Smältpunkt: 48…50 °C
Densitet: 1,354
Storage Temperature: Omgivande
MDL Number: MFCD03790877
CAS nummer: 227617-70-1