1-Hexyl-3-methylimidazolium hexafluorophosphate ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)
H27272.06 H27272.18
H27272.06EA 3325 SEK
H27272.06 H27272.18
1-Hexyl-3-methylimidazolium hexafluorophosphate ≥99%
1-Hexyl-3-methylimidazolium hexafluorophosphate
Purity: 99% CAS No.: 304680-35-1 Molecular Formula: C10H19F6N2P Formula Weight: 312.24 Refractive Index: 1.419
Formel: C₁₀H₁₉F₆N₂P
MW: 312,24 g/mol
Densitet: 1,28 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD03093296
CAS nummer: 304680-35-1