1-Butyl-2,3-dimethylimidazolium chloride ≥99%
Leverantör: Thermo Scientific
Synonyms: 1-Butyl-2,3-dimethyl-3-imidazolium chloride
H27270.06 H27270.18
H27270.06EA 1075 SEK
H27270.06 H27270.18
1-Butyl-2,3-dimethylimidazolium chloride ≥99%
1-Butyl-2,3-dimethylimidazolium chloride
Purity: 99% CAS No.: 98892-75-2 Molecular Formula: C9H17ClN2 Formula Weight: 188.69 Melting Point: 89.5-93.2° MDL No.: MFCD03093291
Formel: C₉H₁₇ClN₂
MW: 188,7 g/mol
Smältpunkt: 89,5…93.2 °C
Storage Temperature: Omgivande
MDL Number: MFCD03093291
CAS nummer: 98892-75-2