1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate ≥99%
Leverantör: Thermo Scientific
Synonyms: 1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate, [BDiMIM] [BF4]
H27141.06 H27141.18
H27141.06EA 1225 SEK
H27141.06 H27141.18
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate ≥99%
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Purity: 99% CAS No.: 402846-78-0 Molecular Formula: C9H17F4N2B Formula Weight: 240.05 MDL No.: MFCD03427618
Formel: C₉H₁₇BF₄N₂
MW: 240,05 g/mol
Densitet: 1,198
Storage Temperature: Omgivande
MDL Number: MFCD03427618
CAS nummer: 402846-78-0