2-Amino-2-ethyl-1,3-propanediol 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Amino-2-ethylpropane-1,3-diol
B24509.22 B24509.36
B24509.22EA 945 SEK
B24509.22 B24509.36
2-Amino-2-ethyl-1,3-propanediol 97%
2-Amino-2-ethyl-1,3-propanediol
Purity: 97% CAS No.: 115-70-8 Molecular Formula: C5H13NO2 Formula Weight: 119.16 Flash Point: 110°(230°F) Refractive Index: 1.4880 Boiling Point: 152-153°/10mm Merck Reference: 14,439 MDL No.: MFCD00004680
Formel: C₅H₁₃NO₂
MW: 119,16 g/mol
Kokpunkt: 152…153 °C (10 mmHg)
Smältpunkt: 39 °C
Densitet: 1,099
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004680
CAS nummer: 115-70-8
EINECS: 204-101-2
Merck Index: 14,439