4-Bromo-2-fluorobenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
B24352.06 B24352.14
B24352.06EA 2350 SEK
B24352.06 B24352.14
4-Bromo-2-fluorobenzenesulphonyl chloride ≥98%
4-Bromo-2-fluorobenzenesulphonyl chloride
Purity: 98% CAS No.: 216159-03-4 Molecular Formula: C6H3BrClFO2S Formula Weight: 273.51 Melting Point: 63-66° Boiling Point: 84°/0.5mm UN No.: 3261 MDL No.: MFCD01090993
Formel: C₆H₃BrClFO₂S
MW: 273,51 g/mol
Kokpunkt: 84 °C (0,5 mmHg)
Smältpunkt: 63…66 °C
Storage Temperature: Omgivande
MDL Number: MFCD01090993
CAS nummer: 216159-03-4
UN: 3261
ADR: 8,II