Ethyl hydrogen adipate ≥98%
Leverantör: Thermo Scientific
Synonyms: Adipic acid monoethyl ester, Adipic acid ethyl ester, 6-Ethoxy-6-oxohexanoic acid, 1,6-Hexanedioic acid monoethyl ester, Monoethyl adipate, 1,6-Hexanedioic acid ethyl ester
B24006.06 B24006.14
B24006.06EA 710 SEK
B24006.06 B24006.14
Ethyl hydrogen adipate ≥98%
Ethyl hydrogen adipate
Purity: 98% CAS No.: 626-86-8 Molecular Formula: HO2C(CH2)4CO2CH2CH3 Formula Weight: 174.20 Flash Point: 110°(230°F) Refractive Index: 1.4390 Melting Point: 28-29° Boiling Point: 179-181° dec. MDL No.: MFCD00004419
Formel: C₈H₁₄O₄
MW: 174,2 g/mol
Kokpunkt: 179…181 °C (dec.)
Smältpunkt: 28…29 °C
Densitet: 0,980
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004419
CAS nummer: 626-86-8
EINECS: 210-966-7