2,4-Bis(trifluoromethyl)benzyl bromide ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-(Bromomethyl)-2,4-bis(trifluoromethyl)benzene
B23910.03 B23910.06
B23910.03EA 1175 SEK
B23910.03 B23910.06
2,4-Bis(trifluoromethyl)benzyl bromide ≥97%
2,4-Bis(trifluoromethyl)benzyl bromide
Purity: 97% CAS No.: 140690-56-8 Molecular Formula: C9H5BrF6 Formula Weight: 307.04 Flash Point: 92°(197°F) Refractive Index: 1.4511 Boiling Point: 112°/0.2mm Beilstein Reference: 7995492 UN No.: 3265 MDL No.: MFCD00010306
Formel: C₉H₅BrF₆
MW: 307.03 g/mol
Kokpunkt: 112 °C (0.2 mmHg)
Densitet: 1.637
Flampunkt: 92 °C (197 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00010306
CAS nummer: 140690-56-8
UN: 3265
ADR: 8 II