2-Methylfuran ≥98% stabilised
Leverantör: Thermo Scientific
Fara
Stabilization: Stabilised med BHT/Ionol 150-400 ppm
B23692.AK B23692.AU
B23692.AKEA 1050 SEK
B23692.AK B23692.AU
2-Methylfuran ≥98% stabilised
2-Methylfuran
Stabilized
Purity: 98+% CAS No.: 534-22-5 Molecular Formula: C5H6O Formula Weight: 82.10 Flash Point: -30°(-22°F) Refractive Index: 1.4330 Melting Point: -89° Boiling Point: 63-65° Beilstein Reference: 103733 UN No.: 2301 MDL No.: MFCD00003248
Formel: C₅H₆O
MW: 82,1 g/mol
Kokpunkt: 63…65 °C
Smältpunkt: –89 °C
Densitet: 0,910
Flampunkt: –30 °C (–22 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003248
CAS nummer: 534-22-5
EINECS: 208-594-5
UN: 2301
ADR: 3,II