trans-1 ,2-diaminocyklohexan-N,N,N',N'-tetraacetic syra monohydrat 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Titriplex® IV
B22928.14 B22928.22
B22928.14EA 1350 SEK
B22928.14 B22928.22
trans-1 ,2-diaminocyklohexan-N,N,N',N'-tetraacetic syra monohydrat 98%
trans-1 ,2-diaminocyklohexan-N,N,N',N'-tetraacetic syra monohydrat
Purity: 98% CAS No.: 125572-95-4 Molecular Formula: C14H22N2O8.H2O Formula Weight: 364.35 (346.34anhy) Melting Point: ca 220° dec. Beilstein Reference: 2226756
Formel: C₁₄H₂₂N₂O₈·1H₂O
MW: 364,35 g/mol
Smältpunkt: ∼ 220 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00066429
CAS nummer: 125572-95-4
EINECS: 236-308-9