5-Hexen-2-one ≥98%
Leverantör: Thermo Fisher Scientific
Varning
B22505.06 B22505.14
B22505.06EA 745 SEK
B22505.06 B22505.14
5-Hexen-2-one ≥98%
5-Hexen-2-one
Purity: 98+% CAS No.: 109-49-9 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 23°(73°F) Refractive Index: 1.4200 Odour: Like ketone Boiling Point: 127-128° Beilstein Reference: 1699137 UN No.: 1224 MDL No.: MFCD00008793
Formel: C₆H₁₀O
Kokpunkt: 127…128 °C
Densitet: 0,847
Flampunkt: 23 °C (73 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008793
CAS nummer: 109-49-9
EINECS: 203-675-1
UN: 1224
ADR: 3,III