Tris(trimethylsilyl)silane ≥97%
Leverantör: Thermo Fisher Scientific
Varning
B22457.06 B22457.03 B22457.14
B22457.06EA 2800 SEK
B22457.06 B22457.03 B22457.14
Tris(trimethylsilyl)silane ≥97%
Tris(trimethylsilyl)silane
Purity: 97% CAS No.: 1873-77-4 Molecular Formula: [(CH3)3Si]3SiH Formula Weight: 248.67 Flash Point: 55°(131°F) Refractive Index: 1.4900 Boiling Point: 82-84°/12mm Beilstein Reference: 1923953 Merck Reference: 14,9757 UN No.: 1993 MDL No.: MFCD00077893
Formel: C₉H₂₈Si₄
Kokpunkt: 82…84 °C (12 mmHg)
Densitet: 0,806
Flampunkt: 55 °C (131 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00077893
CAS nummer: 1873-77-4
ADR: 3,III
Merck Index: 14,9757