2-(4-jodofenyl)-3-(4-nitrofenyl)-5-fenyltetrazoliumklorid 95%
Leverantör: Thermo Scientific
Varning
Synonyms: Iodonitrotetrazolium chloride, p-Iodonitrotetrazolium violet, INT, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazol-3-ium chloride, Iodonitrotetrazolium violet, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyltetrazolium chloride
B22301.03 B22301.06
B22301.03EA 1350 SEK
B22301.03 B22301.06
2-(4-jodofenyl)-3-(4-nitrofenyl)-5-fenyltetrazoliumklorid 95%
2-(4-jodofenyl)-3-(4-nitrofenyl)-5-fenyltetrazoliumklorid
Purity: 95% CAS No.: 146-68-9 Molecular Formula: C19H13ClIN5O2.xH2O Formula Weight: 505.70(anhy) Melting Point: ca 231° dec. Beilstein Reference: 4093224
Formel: C₁₉H₁₃ClIN₅O₂
MW: 505,7 g/mol
Smältpunkt: ∼ 231 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00011961
CAS nummer: 146-68-9
EINECS: 205-676-2