Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid)
Leverantör: Thermo Fisher Scientific
Varning
B21674.09 B21674.18
B21674.09EA 650 SEK
B21674.09 B21674.18
Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid)
Safranine 0 (3,7-diamino-2,8-dimetyl-5-fenylfenaziniumklorid)
CAS No.: 477-73-6 Molecular Formula: C20H19ClN4 Formula Weight: 350.85 Odour: Odourless Beilstein Reference: 3924099 MDL No.: MFCD00011759
Formel: C₂₀H₁₉ClN₄
Densitet: 1,447 g/cm³ (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011759
CAS nummer: 477-73-6
EINECS: 207-518-8