2-Amino-5-nitrothiazole 97%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine
B21335.14
B21335.22
B21335.14EA
720
SEK
B21335.14
B21335.22
2-Amino-5-nitrothiazole 97%
2-Amino-5-nitrothiazole
Purity: 97%
CAS No.: 121-66-4
Molecular Formula: C3H3N3O2S
Formula Weight: 145.14
Melting Point: ca 202° dec.
Beilstein Reference: 126797
Merck Reference: 14,457
MDL No.: MFCD00005326
Formel:
C₃H₃N₃O₂S MW: 145,14 g/mol Smältpunkt: ∼ 202 °C (dec.) Storage Temperature: Omgivande |
MDL Number:
MFCD00005326 CAS nummer: 121-66-4 EINECS: 204-490-9 Merck Index: 14,457 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...