1,3-Dimethylurea ≥98%
Leverantör: Thermo Scientific
Synonyms: N,N'-Dimethylurea
B21329.0B B21329.30 B21329.36
B21329.0BEA 1350 SEK
B21329.0B B21329.30 B21329.36
1,3-Dimethylurea ≥98%
1,3-Dimethylurea
Purity: 98% CAS No.: 96-31-1 Molecular Formula: (CH3NH)2C=O Formula Weight: 88.11 Flash Point: 157°(314°F) Melting Point: 100-104° Boiling Point: 268-270° dec. Beilstein Reference: 1740672 MDL No.: MFCD00008286
Formel: C₃H₈N₂O
MW: 88,11 g/mol
Kokpunkt: 268…270 °C (dec.)
Smältpunkt: 100…104 °C
Densitet: 1,140
Flampunkt: 157 °C (314 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008286
CAS nummer: 96-31-1
EINECS: 202-498-7