Sodium deoxycholate monohydrate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Sodium 3alpha,12alpha-dihydroxy-5beta-cholanoate monohydrate, Deoxycholic acid sodium salt monohydrate, 3α,12α-Dihydroxycholan-24-oic acid sodium salt monohydrate, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt monohydrate, Sodium 3α,12α-dihydroxy-5β-cholanoate monohydrate, 3α,12α-Dihydroxy-5β-cholanic acid sodium salt monohydrate, 3α,12α-Dihydroxycholanic acid sodium salt monohydrate
B20759.14 B20759.22 B20759.36
B20759.14EA 1350 SEK
B20759.14 B20759.22 B20759.36
Sodium deoxycholate monohydrate ≥98%
Sodium deoxycholate monohydrate
Purity: 98% CAS No.: 145224-92-6 Molecular Formula: C24H39NaO4.H2O Formula Weight: 432.58 (414.58anhy) Melting Point: 300° Beilstein Reference: 3643164 Merck Reference: 14,2899 Optical Rotation: +44° (c=2 in water) MDL No.: MFCD00150750
Formel: C₂₄H₃₉NaO₄·1H₂O
MW: 432,58 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00150750
CAS nummer: 145224-92-6
EINECS: 206-132-7
Merck Index: 14,2899