N,N,N',N'-Tetramethyl-1,3-propanediamine ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: N,N,N',N'-Tetramethyl-1,3-diaminopropane, TMPDA, N1,N1,N3,N3-Tetramethylpropane-1,3-diamine, 1,3-Bis(dimethylamino)propane, N,N,N',N'-Tetramethyltrimethylenediamine
A19500.09 A19500.18 A19500.30
A19500.09EA 730 SEK
A19500.09 A19500.18 A19500.30
N,N,N',N'-Tetramethyl-1,3-propanediamine ≥98%
1,3-Bis(dimethylamino)propane
Purity: 99% CAS No.: 110-95-2 Molecular Formula: (CH3)2N(CH2)3N(CH3)2 Formula Weight: 130.24 Flash Point: 31°(87°F) Refractive Index: 1.4234 Boiling Point: 145-146° Beilstein Reference: 1734176 UN No.: 2733 MDL No.: MFCD00008337
Formel: C₇H₁₈N₂
Kokpunkt: 145…146 °C
Densitet: 0,820
Flampunkt: 31 °C (87 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008337
CAS nummer: 110-95-2
EINECS: 203-818-8
UN: 2734
ADR: 8,II