Dinatriumhydrogenarsenat heptahydrat ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Sodium hydrogen arsenate heptahydrate, Sodium arsenate dibasic heptahydrate, Arsenic Acid, Sodium Salt Heptahydrate. Sodium Salt
A18275.22 A18275.36
A18275.22EA 2400 SEK
A18275.22 A18275.36
Dinatriumhydrogenarsenat heptahydrat ≥98%
Dinatriumhydrogenarsenat heptahydrat
Purity: 98% CAS No.: 10048-95-0 Molecular Formula: Na2HAsO4.7H2O Formula Weight: 312.01 (185.94anhy) Odour: Odourless Melting Point: 130° dec. UN No.: 1685 MDL No.: MFCD00149179
Formel: HAsNa₂O₄·7H₂O
Smältpunkt: 130 °C (dec.)
Densitet: 1,88
Storage Temperature: Omgivande
MDL Number: MFCD00149179
CAS nummer: 10048-95-0
EINECS: 231-902-4
UN: 1685
ADR: 6.1,II