6-Methyl-2-thiouracil ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin
A17982.18 A17982.30 A17982.A1
A17982.18EA 520 SEK
A17982.18 A17982.30 A17982.A1
6-Methyl-2-thiouracil ≥98%
6-Methyl-2-thiouracil
Purity: 98% CAS No.: 56-04-2 Molecular Formula: C5H6N2OS Formula Weight: 142.18 Melting Point: ca 329° dec. Beilstein Reference: 115648 Merck Reference: 14,6128 MDL No.: MFCD00006040
Formel: C₅H₆N₂OS
MW: 142,18 g/mol
Kokpunkt: 351 °C (1013 hPa)
Smältpunkt: ∼ 329 °C (dec.)
Densitet: 1,519 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006040
CAS nummer: 56-04-2
EINECS: 200-252-3
Merck Index: 14,6128