Tetraklorftalsyraanhydrid ≥98%
Leverantör: Thermo Scientific
|
Fara
|
A17404.0E
A17404.0C
A17404.36
A17404.0EEA
7375
SEK
A17404.0E
A17404.0C
A17404.36
Tetraklorftalsyraanhydrid ≥98%
Tetraklorftalsyraanhydrid
Purity: 98%
CAS No.: 117-08-8
Molecular Formula: C8Cl4O3
Formula Weight: 285.90
Flash Point: 362°(683°F)
Odour: Weak
Boiling Point: 349-354°
Beilstein Reference: 211560
UN No.: 3077
MDL No.: MFCD00005920
Formel:
C₈Cl₄O₃ MW: 285,9 g/mol Kokpunkt: 349…354 °C Smältpunkt: 254…258 °C Densitet: 1,52 g/cm³ (275 °C) Flampunkt: 362 °C (683 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00005920 CAS nummer: 117-08-8 EINECS: 204-171-4 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...