Benzyltrimethylammonium bromide ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium bromide
A16840.14 A16840.22
A16840.14EA 550 SEK
A16840.14 A16840.22
Benzyltrimethylammonium bromide ≥98%
Benzyltrimethylammonium bromide
Purity: 98+% CAS No.: 5350-41-4 Molecular Formula: C6H5CH2N(CH3)3Br Formula Weight: 230.15 Melting Point: 228-232° Beilstein Reference: 3917251
Formel: C₁₀H₁₆BrN
MW: 230,15 g/mol
Smältpunkt: 228…232 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011780
CAS nummer: 5350-41-4
EINECS: 226-320-2