4,4'-Oxidianilin 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: p-Aminophenyl ether, 4,4'-Oxydianiline, 4,4-Oxydianiline, Bis(4-aminophenyl) ether, 4-Aminophenyl ether
A16815.22 A16815.18 A16815.36
A16815.22EA 1425 SEK
A16815.22 A16815.18 A16815.36
4,4'-Oxidianilin 98%
4,4'-Oxidianilin
Purity: 98% CAS No.: 101-80-4 Molecular Formula: C12H12N2O Formula Weight: 200.24 Flash Point: 218°(424°F) Melting Point: 189-192° Boiling Point: 190°/0.1mm Beilstein Reference: 475735 UN No.: 3077 MDL No.: MFCD00007863
Formel: C₁₂H₁₂N₂O
MW: 200.24 g/mol
Kokpunkt: 190 °C (0.1 mmHg)
Smältpunkt: 189…192 °C
Flampunkt: 218 °C (424 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007863
CAS nummer: 101-80-4
EINECS: 202-977-0
ADR: 9 III