Benzyltrimethylammonium chloride ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride
A16759.0E A16759.22 A16759.36
A16759.0EEA 8675 SEK
A16759.0E A16759.22 A16759.36
Benzyltrimethylammonium chloride ≥97%
Benzyltrimethylammonium chloride
Purity: 97% CAS No.: 56-93-9 Molecular Formula: C6H5CH2N(CH3)3Cl Formula Weight: 185.70 Refractive Index: 1.4440 Odour: Odourless pH: 6-8 Melting Point: ca 238° dec. Beilstein Reference: 3917255 MDL No.: MFCD00011782
Formel: C₁₀H₁₆ClN
MW: 185,7 g/mol
Smältpunkt: ∼ 238 °C (dec.)
Densitet: 1,07 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011782
CAS nummer: 56-93-9
EINECS: 200-300-3
UN: 2811
ADR: 6.1,III