1,3-Dimethyl-2-imidazolidinone ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,3-Dimethylimidazolidin-2-one
A16001.22 A16001.14 A16001.36
A16001.22EA 1250 SEK
A16001.22 A16001.14 A16001.36
1,3-Dimethyl-2-imidazolidinone ≥98%
1,3-Dimethyl-2-imidazolidinone
Purity: 98% CAS No.: 80-73-9 Molecular Formula: C5H10N2O Formula Weight: 114.15 Flash Point: 93°(199°F) Refractive Index: 1.4720 Melting Point: 8° Boiling Point: 224-226° Beilstein Reference: 108808 Merck Reference: 14,3249 UN No.: 2810 MDL No.: MFCD00003188
Formel: C₅H₁₀N₂O
MW: 114,15 g/mol
Kokpunkt: 224…226 °C
Smältpunkt: 8 °C
Densitet: 1,058
Flampunkt: 93 °C (199 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003188
CAS nummer: 80-73-9
EINECS: 201-304-8
UN: 2810
ADR: 6.1,III
Merck Index: 14,3249