2-Chlorobenzyl mercaptan ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-Chlorobenzenemethanethiol, o-Chlorotoluene-α-thiol
A15826.06 A15826.14 A15826.22
A15826.06EA 620 SEK
A15826.06 A15826.14 A15826.22
2-Chlorobenzyl mercaptan ≥97%
2-Chlorobenzyl mercaptan
Purity: 97% CAS No.: 39718-00-8 Molecular Formula: C7H7ClS Formula Weight: 158.65 Flash Point: 69°(156°F) Refractive Index: 1.5910 Boiling Point: 120-122°/25mm Beilstein Reference: 2042183 UN No.: 2810 MDL No.: MFCD00004868
Formel: C₇H₇ClS
Kokpunkt: 120…122 °C (25 mmHg)
Densitet: 1,220
Flampunkt: 69 °C (156 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004868
CAS nummer: 39718-00-8
EINECS: 254-602-5
UN: 2810
ADR: 6.1,III