N-(3-Bromopropyl)phthalimide ≥98%
Leverantör: Thermo Scientific
A15587.14 A15587.22 A15587.36
A15587.14EA 805 SEK
A15587.14 A15587.22 A15587.36
N-(3-Bromopropyl)phthalimide ≥98%
N-(3-Bromopropyl)phthalimide
Purity: 98% CAS No.: 5460-29-7 Molecular Formula: C11H10BrNO2 Formula Weight: 268.12 Odour: Odourless Melting Point: 72-75° Beilstein Reference: 157547 MDL No.: MFCD00005904
Formel: C₁₁H₁₀BrNO₂
MW: 268,11 g/mol
Kokpunkt: 433 °C (1013 hPa)
Smältpunkt: 72…75 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005904
CAS nummer: 5460-29-7
EINECS: 226-738-5