Methyl cyanoacetate ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 2-cyanoacetate
A15555.0B A15555.22 A15555.30 A15555.36
A15555.0BEA 2800 SEK
A15555.0B A15555.22 A15555.30 A15555.36
Methyl cyanoacetate ≥99%
Methyl cyanoacetate
Purity: 99% CAS No.: 105-34-0 Molecular Formula: NCCH2CO2CH3 Formula Weight: 99.09 Flash Point: 110°(230°F) Refractive Index: 1.4185 Odour: Mild Melting Point: -23° Boiling Point: 204° Beilstein Reference: 773945 MDL No.: MFCD00001939
Formel: C₄H₅NO₂
MW: 99,09 g/mol
Kokpunkt: 204 °C
Smältpunkt: –23 °C
Densitet: 1,127
Flampunkt: 110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001939
CAS nummer: 105-34-0
EINECS: 203-288-8