Ethyl levulinate ≥98%
Leverantör: Thermo Scientific
Synonyms: Ethyl 4-oxopentanoate, 4-Oxovaleric acid ethyl ester, Ethyl 4-oxovalerate, 4-Ketovaleric acid ethyl ester, 4-Oxopentanoic acid ethyl ester, Levulinic acid ethyl ester
A15001.18 A15001.22 A15001.36
A15001.18EA 585 SEK
A15001.18 A15001.22 A15001.36
Ethyl levulinate ≥98%
Ethyl levulinate
Purity: 98% CAS No.: 539-88-8 Molecular Formula: C7H12O3 Formula Weight: 144.17 Flash Point: 94°(201°F) Refractive Index: 1.4220 Boiling Point: 203-205° Beilstein Reference: 507641 Merck Reference: 14,3819 MDL No.: MFCD00009209
Formel: C₇H₁₂O₃
MW: 144,17 g/mol
Kokpunkt: 203…205 °C
Densitet: 1,014
Flampunkt: 94 °C (201 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009209
CAS nummer: 539-88-8
EINECS: 208-728-2
Merck Index: 14,3819