Thiophene-3-carbaldehyde 96%
Leverantör: Thermo Scientific
Varning
Synonyms: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde
A14628.09 A14628.18
A14628.09EA 1525 SEK
A14628.09 A14628.18
Thiophene-3-carbaldehyde 96%
Thiophene-3-carbaldehyde
Purity: 96% CAS No.: 498-62-4 Molecular Formula: C5H4OS Formula Weight: 112.15 Storage Temperature: 2°C to 8°C Flash Point: 64°(147°F) Refractive Index: 1.5840 Boiling Point: 194-196° Beilstein Reference: 105889 MDL No.: MFCD00005466
Formel: C₅H₄OS
MW: 112,15 g/mol
Kokpunkt: 194…196 °C
Smältpunkt: –30 °C
Densitet: 1,28
Flampunkt: 64 °C (147 °F)
MDL Number: MFCD00005466
CAS nummer: 498-62-4
EINECS: 207-865-5