Thiophene-2-carbaldehyde ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde
A14344.0B A14344.22 A14344.36
A14344.0BEA 8425 SEK
A14344.0B A14344.22 A14344.36
Thiophene-2-carbaldehyde ≥98%
Thiophene-2-carbaldehyde
Purity: 98+% CAS No.: 98-03-3 Molecular Formula: C5H4OS Formula Weight: 112.15 Storage Temperature: 2°C to 8°C Flash Point: 78°(172°F) Refractive Index: 1.5900 Boiling Point: 196-198° Beilstein Reference: 105819 MDL No.: MFCD00005429
Formel: C₅H₄OS
MW: 112,15 g/mol
Kokpunkt: 196…198 °C
Smältpunkt: ? 10 °C
Densitet: 1,220
Flampunkt: 78 °C (172 °F)
MDL Number: MFCD00005429
CAS nummer: 98-03-3
EINECS: 202-629-8