3-Nitrophthalic acid 95%
Leverantör: Thermo Fisher Scientific
Varning
A14281.22 A14281.30 A14281.36
A14281.22EA 1125 SEK
A14281.22 A14281.30 A14281.36
3-Nitrophthalic acid 95%
Purity: 95% CAS No.: 603-11-2 Molecular Formula: C8H5NO6 Formula Weight: 211.13 Melting Point: ca 214° dec. Beilstein Reference: 2054269
Formel: C₈H₅NO₆
Kokpunkt: 441 °C (1013 hPa)
Smältpunkt: ∼ 214 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007138
CAS nummer: 603-11-2
EINECS: 210-030-8