Bis(2-kloretyl)eter ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2,2'-Diklordietyleter
A14234.0B A14234.18 A14234.30
A14234.0BEA 5050 SEK
A14234.0B A14234.18 A14234.30
Bis(2-kloretyl)eter ≥99%
Bis(2-kloretyl)eter
Purity: 99% CAS No.: 111-44-4 Molecular Formula: C4H8Cl2O Formula Weight: 143.01 Flash Point: 55°(131°F) Refractive Index: 1.4570 Odour: Pungent Melting Point: -50° Boiling Point: 178° Beilstein Reference: 605317 Merck Reference: 14,3066 UN No.: 1916 MDL No.: MFCD00000975
Formel: C₄H₈Cl₂O
MW: 143.01 g/mol
Kokpunkt: 178 °C
Smältpunkt: –50 °C
Densitet: 1.219
Flampunkt: 55 °C (131 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000975
CAS nummer: 111-44-4
EINECS: 203-870-1
UN: 1916
ADR: 6.1 II
Merck Index: 13,03091