(Benzyl)triphenylphosphonium chloride ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Benzyltriphenylphosphonium chloride
A14034.14 A14034.22 A14034.36
A14034.14EA 765 SEK
A14034.14 A14034.22 A14034.36
(Benzyl)triphenylphosphonium chloride ≥99%
(Benzyl)triphenylphosphonium chloride
Purity: 99% CAS No.: 1100-88-5 Molecular Formula: C6H5CH2P(C6H5)3Cl Formula Weight: 388.88 Flash Point: 300°(572°F) Odour: Odourless Beilstein Reference: 3599868 UN No.: 3464 MDL No.: MFCD00011913
Formel: C₂₅H₂₂ClP
MW: 388,88 g/mol
Smältpunkt: ∼ 330 °C (dec.)
Densitet: 1,18 g/cm³ (20 °C)
Flampunkt: >300 °C (572 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011913
CAS nummer: 1100-88-5
EINECS: 214-154-3
UN: 3464
ADR: 6.1,II