3-Acetyl-2,5-dichlorothiophene ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-(2,5-Dichloro-3-thienyl)-1-ethanone
A13867.06 A13867.14 A13867.22
A13867.06EA 1725 SEK
A13867.06 A13867.14 A13867.22
3-Acetyl-2,5-dichlorothiophene ≥98%
3-Acetyl-2,5-dichlorothiophene
Purity: 98% CAS No.: 36157-40-1 Molecular Formula: C6H4Cl2OS Formula Weight: 195.07 Flash Point: 110°(230°F) Melting Point: 37-39° Boiling Point: 120-122°/4mm Beilstein Reference: 121288 MDL No.: MFCD00014522
Formel: C₆H₄Cl₂OS
MW: 195,07 g/mol
Kokpunkt: 120…122 °C (4 mmHg)
Smältpunkt: 37…39 °C
Flampunkt: 110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00014522
CAS nummer: 36157-40-1
EINECS: 252-893-3