3-(4-Methoxyphenyl)propionic acid ≥98%
Leverantör: Thermo Scientific
Synonyms: 3-(4-Methoxyphenyl)propanoic acid
A13655.14 A13655.22
A13655.14EA 1800 SEK
A13655.14 A13655.22
3-(4-Methoxyphenyl)propionic acid ≥98%
3-(4-Methoxyphenyl)propionic acid
Purity: 98% CAS No.: 1929-29-9 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 102-106° Beilstein Reference: 1953326
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Kokpunkt: 319 °C (1013 hPa)
Smältpunkt: 102…106 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002777
CAS nummer: 1929-29-9
EINECS: 217-678-0