Bentonit, pulver NF
Leverantör: Spectrum Chemical
Synonyms: Montmorillonite, Bentonite
BE125-12KG BE125-2.5KG BE125-22.5KGB BE125-500GM
SPCMBE125-12KGEA 19300 SEK
SPCMBE125-12KG SPCMBE125-2.5KG SPCMBE125-22.5KGB SPCMBE125-500GM
Bentonit, pulver NF
Bentonite
Bentonite, Powder, NF is used as suspending and emulsifying agent.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Densitet: 2.5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00130611
CAS nummer: 1302-78-9
EINECS: 215-108-5